Merck
  • Replacing Fish Meal with Defatted Insect Meal (Yellow Mealworm Tenebrio molitor) Improves the Growth and Immunity of Pacific White Shrimp (Litopenaeus vannamei).
Replacing Fish Meal with Defatted Insect Meal (Yellow Mealworm Tenebrio molitor) Improves the Growth and Immunity of Pacific White Shrimp (Litopenaeus vannamei).
Animals : an open access journal from MDPI (2019-05-24)
Constant Motte, Alfredo Rios, Thomas Lefebvre, Hong Do, Morgane Henry, Orapint Jintasataporn
ZUSAMMENFASSUNG
Recently, ecological and economic issues have affected fish meal (FM) supply, the main source of protein for shrimp. This triggered a search for alternative dietary protein sources for shrimp production. We studied the consequences of replacing FM with a defatted insect meal, ŸnMealTM (YM), comprised of yellow mealworm (Tenebrio molitor). Growth and immune parameters of juvenile Pacific white shrimp (Litopenaeus vannanmei) were compared after an eight-week feeding trial. Shrimp were kept in aquaria with densities of 60 and 40 shrimp/m2 and fed one of five diets in which a proportion of FM was replaced by YM. All diets were isoproteic, isoenergetic, and balanced in lysine and methionine. After the feeding trial, shrimp were challenged with pathogenic bacteria (Vibrio parahaemolyticus). Growth and feed conversion parameters improved when YM was included in shrimp diets; with the highest weight gain and best food conversion ratio (FCR) achieved when 50% of FM was replaced by YM versus the control diet that contained no YM (initial weight: 1.60 g/shrimp; growth: 5.27 vs. 3.94 g/shrimp; FCR 1.20 vs. 1.59). In challenged shrimp, mortality rates were significantly less among groups that received YM, with a 76.9% lower mortality rate in the 50% FM replacement group versus the control.