Merck
  • The effect of a short-term low-carbohydrate, high-fat diet with or without postmeal walks on glycemic control and inflammation in type 2 diabetes: a randomized trial.
The effect of a short-term low-carbohydrate, high-fat diet with or without postmeal walks on glycemic control and inflammation in type 2 diabetes: a randomized trial.
American journal of physiology. Regulatory, integrative and comparative physiology (2018-10-12)
Étienne Myette-Côté, Cody Durrer, Helena Neudorf, Tyler D Bammert, José Diego Botezelli, James D Johnson, Christopher A DeSouza, Jonathan P Little
ZUSAMMENFASSUNG
Lowering carbohydrate consumption effectively lowers glucose, but impacts on inflammation are unclear. The objectives of this study were to: 1) determine whether reducing hyperglycemia by following a low-carbohydrate, high-fat (LC) diet could lower markers of innate immune cell activation in type 2 diabetes (T2D) and 2) examine if the combination of an LC diet with strategically timed postmeal walking was superior to an LC diet alone. Participants with T2D ( n = 11) completed a randomized crossover study involving three 4-day diet interventions: 1) low-fat low-glycemic index (GL), 2) and 3) LC with 15-min postmeal walks (LC+Ex). Four-day mean glucose was significantly lower in the LC+Ex group as compared with LC (-5%, P < 0.05), whereas both LC+Ex (-16%, P < 0.001) and LC (-12%, P < 0.001) conditions were lower than GL. A significant main effect of time was observed for peripheral blood mononuclear cells phosphorylated c-Jun N-terminal kinase ( P < 0.001), with decreases in all three conditions (GL: -32%, LC: -45%, and LC+Ex: -44%). A significant condition by time interaction was observed for monocyte microparticles ( P = 0.040) with a significant decrease in GL (-76%, P = 0.035) and a tendency for a reduction in LC (-70%, P = 0.064), whereas there was no significant change in LC+Ex (0.5%, P = 0.990). Both LC (-27%, P = 0.001) and LC+Ex (-35%, P = 0.005) also led to significant reductions in circulating proinsulin. An LC diet improved 4-day glycemic control and fasting proinsulin levels when compared with GL, with added glucose-lowering benefits when LC was combined with postmeal walking.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete, Mini, EDTA-freier Protease-Inhibitor-Cocktail, Tablets provided in EASYpacks