Merck
Terc is dispensable for most of the short-term HPV16 oncogene-mediated phenotypes in mice.
PloS one (2018-04-27)
Charis Achilleos, Stella Michael, Katerina Strati
ZUSAMMENFASSUNG
High-risk human papillomaviruses (HPVs) have been shown in vitro to impinge on telomere homeostasis in a number of ways. However, the in vivo interaction of viruses with the telomere homeostasis apparatus has not been previously explored. Since E6 and E7 are the main viral oncogenes and key for viral replication, we have explored here the short-term phenotypes of the genes in the context of defective telomere homeostasis. We examined the short-term phenotypes of E6 and E7 in a context where the Terc component of the telomerase holoenzyme was knocked out. We determined that Terc was dispensable for most oncogene-mediated phenotypes. Surprisingly, E7-mediated reduction of label retaining cells was found to be in part dependent on the presence of Terc. Under the conditions examined here, there appears to be no compelling evidence Terc is required for most short-term viral oncogene mediated phenotypes. Further studies will elucidate its role in longer-term phenotypes.