Merck
  • The utility of a point-of-care sural nerve conduction device for detection of diabetic polyneuropathy: A cross-sectional study.
The utility of a point-of-care sural nerve conduction device for detection of diabetic polyneuropathy: A cross-sectional study.
Muscle & nerve (2018-12-26)
Mustafa A Kural, Signe T Andersen, Niels T Andersen, Henning Andersen, Morten Charles, Nanna B Finnerup, Troels S Jensen, Hatice Tankisi
ZUSAMMENFASSUNG
Rapid and accessible methods for diagnosing diabetic polyneuropathy (DPN) have been developed, but not validated, in large cohorts of people with diabetes. The performance of a point-of-care device (POCD) was studied in 168 patients with type 2 diabetes, estimating the sensitivity, specificity, positive predictive value (PPV), and negative predictive value (NPV) compared with conventional sural nerve conduction studies (NCS). A POCD amplitude limit of 6 µV increased the sensitivity (96%) and NPV (98%), but decreased the specificity (71%) and PPV (54%) compared with the 4-µV limit, which had values of 78%, 92%, 89%, and 71%, respectively. POCD on both legs showed better performance than on 1 leg. POCD amplitudes and conduction velocities correlated significantly with conventional sural NCS, but POCD values were underestimated compared with NCS. The POCD may be used as a suitable screening tool for detection of DPN. Patients with abnormal and borderline results should undergo conventional NCS. Muscle Nerve 59:187-193, 2019.