Merck
  • Cationic Pd(II)-catalyzed highly enantioselective arylative cyclization of alkyne-tethered enals or enones initiated by carbopalladation of alkynes with arylboronic acids.
Cationic Pd(II)-catalyzed highly enantioselective arylative cyclization of alkyne-tethered enals or enones initiated by carbopalladation of alkynes with arylboronic acids.
Organic letters (2012-03-28)
Kun Shen, Xiuling Han, Xiyan Lu
ZUSAMMENFASSUNG
Cationic Pd(II)-catalyzed enantioselective arylative cyclization of alkyne-tethered enals or enones initiated by carbopalladation of alkynes was developed without the necessity of a redox system.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Bromphenylboronsäure, ≥95.0%
Sigma-Aldrich
4-Chlorphenylborsäure, 95%