Merck
A chrysin analog exhibited strong inhibitory activities against both PGE2 and NO production.
European journal of medicinal chemistry (2011-07-02)
Haiyan Che, Hyun Lim, Hyun Pyo Kim, Haeil Park
ZUSAMMENFASSUNG
A number of 8-substituted chrysin analogs have been prepared and evaluated for their inhibitory activities of cyclooxygenase-2 catalyzed prostaglandin E(2) and iNOS-mediated NO production. Analogs were prepared via Suzuki-Miyaura C-C cross coupling reaction of 8-iodo-5,7-dimethoxyflavone and alkyl/areneboronic acids as a key reaction. Among the analogs tested, 5,7-dihydroxy-8-(pyridine-4-yl)flavone (3d) showed strong biological activities against COX-2 catalyzed PGE(2) and iNOS-mediated NO production from LPS-induced RAW 264.7 cells compared to those of chrysin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methylborsäure, 97%
Sigma-Aldrich
Chrysin, ≥96.5%