Merck
  • Size-Selective Separation of Rare Earth Elements Using Functionalized Mesoporous Silica Materials.
Size-Selective Separation of Rare Earth Elements Using Functionalized Mesoporous Silica Materials.
ACS applied materials & interfaces (2019-05-24)
Yimu Hu, Luis C Misal Castro, Elisabeth Drouin, Justyna Florek, Hanspeter Kählig, Dominic Larivière, Freddy Kleitz, Frédéric-Georges Fontaine
ZUSAMMENFASSUNG
The separation and preconcentration of rare earth elements (REEs) from mineral concentrates in an economically and environmentally sustainable manner are difficult tasks due to their similar physicochemical properties. Herein, a series of tetradentate phenylenedioxy diamide (PDDA) ligands were synthesized and grafted on large-pore three-dimensional KIT-6 mesoporous silica. In solid-phase extraction, the hybrid sorbents enable a size-selective separation of REEs on the basis of the bite angles of the ligands. In particular, smaller REE3+ ions are preferentially extracted by KIT-6-1,2-PDDA, whereas light REEs with larger ionic radius are favored by KIT-6-1,3-PDDA. The exposure of bauxite residue digestion solution containing REEs as well as a number of types of competitive ions (including Th and U) to the sorbents results in selective recovery of target REEs. The possibility of regenerating the mesoporous sorbents through a simple loading-stripping-regeneration process is demonstrated over up to five cycles with no significant loss in REE extraction capacity, suggesting adequate chemical and structural stability of the new sorbent materials.