Merck
  • Novel effects of sphingosylphosphorylcholine on the apoptosis of breast cancer via autophagy/AKT/p38 and JNK signaling.
Novel effects of sphingosylphosphorylcholine on the apoptosis of breast cancer via autophagy/AKT/p38 and JNK signaling.
Journal of cellular physiology (2018-11-30)
Di Ge, Jia Gao, Lina Han, Ying Li, Hong-Hong Liu, Wan-Cheng Yang, Fen Chang, Jing Liu, Mei Yu, Jing Zhao
ZUSAMMENFASSUNG
Sphingosylphosphorylcholine (SPC), an important lipid mediator in blood, inhibits the proliferation and migration of various cancer cells. However, its effect as a cell-specific sphingolipid in breast cancer cells is still unknown. Here, we showed that SPC promoted autophagy and apoptosis in triple-negative breast cancer MDA-MB-231 cells. Autophagy worked as a negative regulator of apoptosis-induced by SPC. Mechanistically, SPC mediated apoptosis via activating c-Jun N-terminal kinase (JNK). Meanwhile, p38MAPK (p38) and protein kinase B (PKB or AKT) signaling pathways were also activated to inhibit apoptosis, suggesting that SPC could evoke multiple signaling pathways to modulate cell apoptosis. In addition, the crosstalk between autophagy, p38, AKT and JNK is that autophagy, p38, and AKT attenuated the JNK. AKT and p38 were in the downstream of autophagy, which is autophagy/AKT/p38 signaling evoked by SPC to antagonize JNK signaling and subsequent apoptosis. Although the pathways that antagonize apoptosis were evoked, the cells eventually reached apoptosis by SPC. Therefore, the combination with pharmacological autophagy inhibitors would be a more effective therapeutic strategy for eliminating breast cancer cells by SPC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
3-Methyladenin, autophagy inhibitor
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)
Sigma-Aldrich
Wortmannin aus Penicillium funiculosum, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder