Merck
  • Edible Coatings Enriched with Essential Oils on Apples Impair the Survival of Bacterial Pathogens through a Simulated Gastrointestinal System.
Edible Coatings Enriched with Essential Oils on Apples Impair the Survival of Bacterial Pathogens through a Simulated Gastrointestinal System.
Foods (Basel, Switzerland) (2019-02-06)
Ana Isabel Vieira, Adriana Guerreiro, Maria Dulce Antunes, Maria da Graça Miguel, Maria Leonor Faleiro
ZUSAMMENFASSUNG
Edible coatings supplemented with essential oil components have been investigated to control spoilage microorganisms. In this study, the survival of Listeria monocytogenes and Salmonella enterica serovar Typhimurium on apples treated with edible coatings based on sodium alginate (2%) (ECs) and supplemented with essential oil components, namely eugenol (Eug) at 0.2% or in combination with 0.1% (v/v) of Eug and citral (Cit) at 0.15% was determined. Both bacterial pathogens were exposed on apples treated with ECs supplemented with Eug or Eug + Cit and challenged with gastrointestinal fluids and their survival was examined. Both pathogens were able to survive on the surface of 'Bravo de Esmolfe' apple. The use of ECs in fresh-cut fruits impaired the survival of both bacterial populations over 72 h at 4 °C. The exposure of the pathogens on apples with ECs supplemented with Eug and Cit and challenged with gastrointestinal fluids significantly reduced their survival. This study evidences that the use of alginate edible coating enriched with Eug or the combination of Eug and Cit can contribute to the safer consumption of minimally processed fruits.