Merck
  • Determination of plutonium isotopes in environmental samples by extraction chromatography with triisooctylamine - polyethylene resin.
Determination of plutonium isotopes in environmental samples by extraction chromatography with triisooctylamine - polyethylene resin.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2019-02-13)
L Popov
ZUSAMMENFASSUNG
Method for determination of plutonium isotopes in various environmental samples is presented. The separation and purification of plutonium is attained by extraction chromatography with triisooctylamine - polyethylene resin in nitric and hydrochloric acid media. Plutonium is measured by alpha-particle spectrometry after electrodeposition on a stainless steel disk. The analytical quality was checked by analyzing reference materials with different matrices from IAEA (Soil-6, IAEA-375, IAEA-384, IAEA-414) and NPL (AL-2009, AL-2010, AL-2011, AL-2013, AB-2014). The major advantages of the method are the low cost of the analysis, high radiochemical yields and high decontamination factors from the matrix elements, natural and man-made radionuclides.