Merck
  • Asymmetric Allylboration of Aldehydes with Pinacol Allylboronates Catalyzed by 1,1'-Spirobiindane-7,7'-diol (SPINOL) Based Phosphoric Acids
Asymmetric Allylboration of Aldehydes with Pinacol Allylboronates Catalyzed by 1,1'-Spirobiindane-7,7'-diol (SPINOL) Based Phosphoric Acids
European Journal of Organic Chemistry (2012)
Xing, C-H.; et al.