Merck
  • Tubulin acetylation accompanies autophagy development induced by different abiotic stimuli in Arabidopsis thaliana.
Tubulin acetylation accompanies autophagy development induced by different abiotic stimuli in Arabidopsis thaliana.
Cell biology international (2017-08-18)
Vira Olenieva, Dmytro Lytvyn, Alla Yemets, Catherine Bergounioux, Yaroslav Blume
ZUSAMMENFASSUNG
Microtubules (MTs) play an important role in the regulation of autophagy development in yeast and animal as well as in plant cells. MTs participate in maturation and traffic of autophagosomes through their dynamic state changes and post-translational modifications of tubulin, namely acetylation. We subjected Arabidopsis thaliana seedlings to metabolic-, salt-, osmotic stresses as well as irradiation of ultraviolet B and investigated the involvement of plant MTs in the development of stress-induced autophagy via tubulin acetylation. For this purpose Arabidopsis thaliana line expressing autophagy-related protein 8 h (atg8h)-GFP was generated to investigate autophagy, applying the level of free GFP as an indicator of autophagy development. Using autophagosome confocal imaging and Western blot analysis of Atg8 post-translational lipidation and synchronous GFP release it was shown that all examined stressful stimuli led to pronounced development of autophagy, particularly in different root tissues. Moreover, autophagy development was accompanied by α-tubulin acetylation under all stressful conditions. Presented data indicate the possible role of the post-translational acetylation of α-tubulin in the mediation of plant stress-induced autophagy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for plant cell and tissue extracts, DMSO solution
Sigma-Aldrich
Monoklonales Anti-Tubulin, acetyliert in Maus hergestellte Antikörper, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-GFP, N-terminal in Kaninchen hergestellte Antikörper, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution