Merck
  • The inhibition of thymidine phosphorylase can reverse acquired 5FU-resistance in gastric cancer cells.
The inhibition of thymidine phosphorylase can reverse acquired 5FU-resistance in gastric cancer cells.
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association (2018-10-03)
Ryutaro Mori, Kazuhiro Yoshida, Manabu Futamura, Tomonari Suetsugu, Kaoru Shizu, Toshiyuki Tanahashi, Yoshihiro Tanaka, Nobuhisha Matsuhashi, Kazuya Yamaguchi
ZUSAMMENFASSUNG
5FU can be converted to its active metabolite fluoro-deoxyuridine monophosphate (FdUMP) through two pathways: the orotate phosphoribosyl transferase-ribonucleotide reductase (OPRT-RR) pathway and the thymidine phosphorylase-thymidine kinase (TP-TK) pathway. We investigated the mechanism underlying 5FU-resistance, focusing on the changes in the 5FU metabolisms. MKN45 and 5FU-resistant MKN45/F2R cells were treated with 5FU or fluoro-deoxyuridine (FdU) in combination with hydroxyurea (HU) or tipiracil (TPI). The amount of FdUMP was determined by the density of the upper band of thymidylate synthase on Western blotting. The MKN45/F2R cells exhibited 5FU resistance (37.1-fold) and showed decreased OPRT and increased TP levels. In both cells, the FdUMP after treatment with 5FU was decreased when RR was inhibited by HU but not when TP was inhibited by TPI. A metabolome analysis revealed the loss of intracellular deoxyribose 1-phosphate (dR1P) in both cells, indicating that FdUMP was synthesized from 5FU only through the OPRT-RR pathway because of the loss of dR1P. After the knockdown of TK, the FdUMP after treatment with FdU was decreased in MKN45 cells. However, it was not changed in MKN45/F2R cells. Furthermore, TP inhibition caused an increase in FdUMP after treatment with 5FU or FdU and reversed the 5FU resistance in MKN45/F2R cells, indicating that FdUMP was reduced through the TP-TK pathway in MKN45/F2R cells. In MKN45/F2R cells, the reduction of FdUMP through the TP-TK pathway caused 5FU resistance, and the inhibition of TP reversed the resistance to 5FU, suggesting that the combination of 5FU and TPI is a promising cancer therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Maus-IgG-Antikörper der Ziege, Peroxidase-konjugiert, H+L, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Peroxidase Conjugated, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Anti-Thymidylate Synthase Antibody, clone TS106, clone TS106, Chemicon®, from mouse