Merck
Cyclized NDGA modifies dynamic α-synuclein monomers preventing aggregation and toxicity.
Scientific reports (2019-03-01)
Malcolm J Daniels, J Brucker Nourse, Hanna Kim, Valerio Sainati, Marco Schiavina, Maria Grazia Murrali, Buyan Pan, John J Ferrie, Conor M Haney, Rani Moons, Neal S Gould, Antonino Natalello, Rita Grandori, Frank Sobott, E James Petersson, Elizabeth Rhoades, Roberta Pierattelli, Isabella Felli, Vladimir N Uversky, Kim A Caldwell, Guy A Caldwell, Edward S Krol, Harry Ischiropoulos
ZUSAMMENFASSUNG
Growing evidence implicates α-synuclein aggregation as a key driver of neurodegeneration in Parkinson's disease (PD) and other neurodegenerative disorders. Herein, the molecular and structural mechanisms of inhibiting α-synuclein aggregation by novel analogs of nordihydroguaiaretic acid (NDGA), a phenolic dibenzenediol lignan, were explored using an array of biochemical and biophysical methodologies. NDGA analogs induced modest, progressive compaction of monomeric α-synuclein, preventing aggregation into amyloid-like fibrils. This conformational remodeling preserved the dynamic adoption of α-helical conformations, which are essential for physiological membrane interactions. Oxidation-dependent NDGA cyclization was required for the interaction with monomeric α-synuclein. NDGA analog-pretreated α-synuclein did not aggregate even without NDGA-analogs in the aggregation mixture. Strikingly, NDGA-pretreated α-synuclein suppressed aggregation of naïve untreated aggregation-competent monomeric α-synuclein. Further, cyclized NDGA reduced α-synuclein-driven neurodegeneration in Caenorhabditis elegans. The cyclized NDGA analogs may serve as a platform for the development of small molecules that stabilize aggregation-resistant α-synuclein monomers without interfering with functional conformations yielding potential therapies for PD and related disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(−)-Epigallokatechingallat, ≥95%