Merck
  • Trans-β-galactosidase activity of pig enzymes embedded in the small intestinal brush border membrane vesicles.
Trans-β-galactosidase activity of pig enzymes embedded in the small intestinal brush border membrane vesicles.
Scientific reports (2019-02-01)
Lesbia Cristina Julio-Gonzalez, Oswaldo Hernandez-Hernandez, F Javier Moreno, Agustín Olano, Maria Luisa Jimeno, Nieves Corzo
ZUSAMMENFASSUNG
This work highlights the utility of brush border membrane vesicles (BBMV) from the pig small intestine as a reliable model for gathering information about the reaction mechanisms involved in the human digestion of dietary carbohydrates. Concretely, the elucidation of the transgalactosylation mechanism of pig BBMV to synthesize prebiotic galacto-oligosaccharides (GOS) is provided, unravelling the catalytic activity of mammalian small intestinal β-galactosidase towards the hydrolysis of GOS. This study reveals that pig BBMV preferably synthesizes GOS linked by β-(1 → 3) bonds, since major tri- and disaccharide were produced by the transfer of a galactose unit to the C-3 of the non-reducing moiety of lactose and to the C-3 of glucose, respectively. Therefore, these results point out that dietary GOS having β-(1 → 3) as predominant glycosidic linkages could be more prone to hydrolysis by mammalian intestinal digestive enzymes as compared to those linked by β-(1 → 2), β-(1 → 4), β-(1 ↔ 1) or β-(1 → 6). Given that these data are the first evidence on the transglycosylation activity of mammalian small intestinal glycosidases, findings contained in this work could be crucial for future studies investigating the structure-small intestinal digestibility relationship of a great variety of available prebiotics, as well as for designing tailored fully non-digestible GOS.