Merck
  • Microbial Community Enhances Biodegradation of Bisphenol A Through Selection of Sphingomonadaceae.
Microbial Community Enhances Biodegradation of Bisphenol A Through Selection of Sphingomonadaceae.
Microbial ecology (2018-09-27)
Seungdae Oh, Donggeon Choi
ZUSAMMENFASSUNG
Bisphenol A (BPA) is a common ingredient in plastic wares and epoxy resins that are essential for our daily life. Despite the obvious benefits, BPA may act as an environmental endocrine disruptor, causing metabolic, reproductive, and/or developmental consequences and diseases in humans and other organisms. Although previous studies have yielded progress toward the microbial breakdown of BPA, the work has primarily been focused on pure cultures rather than complex microbial communities. In this study, we examined microbial communities in bioreactors that control the fate of BPA at various levels (up to 5000 μg L-1). Microbial communities rapidly increased removal rates of 500-5000 μg L-1 BPA from 23-29 to 89-99% during the first 2 weeks of the acclimation period, after which > 90% stable removal rates were maintained over 3 months. Biochemical assays demonstrated that BPA was removed by biodegradation, rather than other abiotic removal routes (e.g., adsorption and volatilization). The 16S rRNA gene-based community analysis revealed that 50-5000 μg L-1 of BPA exposure systematically selected for three Sphingomonadaceae species (Sphingobium, Novosphingobium, and Sphingopyxis). The Sphingomonadaceae-enriched communities acclimated to BPA showed a 7.0-L gVSS-1 day-1 BPA degradation rate constant, which is comparable to that (4.1-6.3) of Sphingomonadaceae isolates and is higher than other potential BPA degraders. Taken together, our results advanced the understanding of how microbial communities acclimate to environmentally relevant levels of BPA, gradually enhancing BPA degradation via selective enrichment of a few Sphingomonadaceae populations with higher BPA metabolic activity.