Merck
  • Activin A regulates the epidermal growth factor receptor promoter by activating the PI3K/SP1 pathway in oral squamous cell carcinoma cells.
Activin A regulates the epidermal growth factor receptor promoter by activating the PI3K/SP1 pathway in oral squamous cell carcinoma cells.
Scientific reports (2019-03-28)
Chi-Neu Tsai, Chia-Lung Tsai, Jui-Shan Yi, Huang-Kai Kao, Yenlin Huang, Chun-I Wang, Yun-Shien Lee, Kai-Ping Chang
ZUSAMMENFASSUNG
Epidermal growth factor receptor (EGFR) and activin A are both overexpressed in oral cavity squamous cell carcinoma (OSCC). We evaluated their clinical correlation and activin A-mediated EGFR regulation in this study. Overexpression of both transcripts/proteins indicated a poorer prognosis in OSCC patients. Knockdown of endogenous INHBA repressed the expression of EGFR and inhibited activin A-mediated canonical Smads, noncanonical phosphorylation of AKT (ser473) (p-AKT ser473) and SP1. Inhibition of PI3K signaling via its inhibitor attenuated p-AKT ser473 and in turn reduced SP1 and EGFR expression in the presence of recombinant activin A (rActivin A) in OSCC cells, as revealed via a luciferase assay and western blotting. However, canonical Smad signaling repressed the EGFR promoter, as revealed by a luciferase assay. The transcription factor SP1, its coactivator CBP/p300, and Smad proteins were recruited to the EGFR proximal promoter following rActivin A treatment, as revealed by chromatin immunoprecipitation (ChIP). Smad2/3/4 dramatically outcompeted SP1 binding to the EGFR proximal promoter following mithramycin A treatment. Activin A activates the PI3K and Smad pathways to compete for binding to overlapping SP1 consensus sequences on the EGFR proximal promoter. Nevertheless, canonical p-Smad2 was largely repressed in OSCC tumor tissues, suggesting that the activin A-mediated noncanonical pathway is essential for the carcinogenesis of OSCC.