- Identification and biosynthesis of 2-(1H-imidazol-5-yl) ethan-1-ol (histaminol) in methanogenic archaea.
Identification and biosynthesis of 2-(1H-imidazol-5-yl) ethan-1-ol (histaminol) in methanogenic archaea.
Microbiology (Reading, England) (2019-02-05)
Rebecca Portugal, Nana Shao, William B Whitman, Kylie D Allen, Robert H White
PMID30714894
ZUSAMMENFASSUNG
Histaminol is a relatively rare metabolite most commonly resulting from histidine metabolism. Here we describe histaminol production and secretion into the culture broth by the methanogen Methanococcus maripaludis S2 as well as a number of other methanogens. This work is the first identification of this compound as a natural product in methanogens. Its biosynthesis from histidine was confirmed by the incorporation of 2H3-histidine into histaminol by growing cells of M. maripaludis S2. Possible functions of this molecule could be cell signaling as observed with histamine in eukaryotes or uptake of metal ions.