Merck
  • Evidence of impacts from DDT in pelican, cormorant, stork, and egret eggs from KwaZulu-Natal, South Africa.
Evidence of impacts from DDT in pelican, cormorant, stork, and egret eggs from KwaZulu-Natal, South Africa.
Chemosphere (2019-03-23)
H Bouwman, Y B Yohannes, S M M Nakayama, K Motohira, M Ishizuka, M S Humphries, V van der Schyff, M du Preez, A Dinkelmann, Y Ikenaka
ZUSAMMENFASSUNG
DDT remains in use for malaria control in South Africa. We quantified DDTs in aquatic bird eggs from the highly biodiverse northern KwaZulu-Natal, a province of South Africa where DDT has been used for more than 80 years for malaria control. Pelican eggs had the highest ΣDDT concentration (7200 ng/g lipid mass; lm), Little Egret eggs had 6900 ΣDDT lm, African Openbill eggs had 3400 ng/g lm ΣDDT, and White-breasted Cormorant had 2400 ng/g lm. All species had non-significantly different mean concentrations of o,p'-DDT, p,p'-DDT, and ΣDDT, but with significant differences for p,p-DDE, o,p'-DDD, p,p'-DDD, %DDT, %DDD, and %lipid. The thinnest pelican eggshell (0.40 mm) had a ΣDDT concentration of 3300 ng/g lm.; the thickest shell (0.96 mm) had the lowest ΣDDT concentration at 29 ng/g lm; a 58% difference. Linear regressions of concentrations with shell thickness for the pelican eggs were significant for p,p'-DDE and p,p'-DDD, indicating risk of reproductive impairment. Compositional profiles indicate different food webs for the different species. DDT concentrations were lower than from another DDT-sprayed locality in South Africa, possible linked to differences in hydrology and rainfall. We conclude that significant ecotoxic threats associated with DDT remain in this area, and possibly threatens birds from less polluted areas. Our findings suggest continued negative human health and environmental impacts from DDT. There is an urgency to move away from DDT as quickly as possible; alternatively, to implement practices that prevent emissions of DDT to the environment while protecting human life.