Merck
Cellular internalization and trafficking of 20 KDa human growth hormone.
General and comparative endocrinology (2018-10-20)
Lan Hai-Nan, Liu Hui-Lin, Zhang Zi-Qi, Luo Gan, Fu Xue-Qi, Zheng Xin
ZUSAMMENFASSUNG
Twenty kilodalton human growth hormone (20K-GH) is the second most abundant GH isoform after the twenty-two kilodalton human growth hormone (22 K-GH) isoform. 20K-GH exhibits similar but not identical physiological activities as that of 22K-GH. The cell behaviour of 22K-GH has been extensively studied, but little or no information has been reported regarding 20K-GH. Here, we focussed on the internalization of 20K-GH. We found that the internalization of 20K-GH is rapid and occurs in a time- and dose-dependent manner. 20K-GH internalization is mediated by GHR. It appears that the internalization of 20K-GH and GHR into the cytoplasm is mediated by clathrin and/or caveolin. The current study indicates that 20K-GH can internalize into the cytoplasm and suggests that the internalized 20K-GH may exhibit different functions from those of 22K-GH.