Merck
3D arrays of microcages by two-photon lithography for spatial organization of living cells.
Lab on a chip (2019-02-07)
Florian Larramendy, Shotaro Yoshida, Daniela Maier, Zoltan Fekete, Shoji Takeuchi, Oliver Paul
ZUSAMMENFASSUNG
This paper addresses a nanoengineering approach to create a fully three-dimensional (3D) network of living cells, providing an advanced solution to in vitro studies on either neuronal networks or artificial organs. The concept of our work relies on stackable scaffolds composed of microcontainers designed and dimensioned to favor the geometrically constrained growth of cells. The container geometry allows cells to communicate in the culture medium and freely grow their projections to form a 3D arrangement of living cells. Scaffolds are fabricated using two-photon polymerization of IP-L 780 photoresist and are coated with collagen. They are stacked by mechanical micromanipulation. Technical details of the proposed nanofabrication scheme and assembly of the modular culture environment are explained. Preliminary in vitro results using PC12 cells have shown that this structure provides a good basis for healthy cell growth for at least 16 days. Our approach is envisioned to provide tailor-made solutions of future 3D cell assemblies for potential applications in drug screening or creating artificial organs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dulbecco Modifiziertes Eagle-Medium – hoher Glucosegehalt, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dulbeccos Phosphatgepufferte Kochsalzlösung, 10×, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Nervenwachstumsfaktor 2.5S aus Maussubmaxillardrüse, NGF-2.5S, lyophilized powder, suitable for cell culture