Merck
D-Aspartate treatment attenuates myelin damage and stimulates myelin repair.
EMBO molecular medicine (2018-12-19)
Valeria de Rosa, Agnese Secondo, Anna Pannaccione, Roselia Ciccone, Luigi Formisano, Natascia Guida, Roberta Crispino, Annalisa Fico, Roman Polishchuk, Antimo D'Aniello, Lucio Annunziato, Francesca Boscia
ZUSAMMENFASSUNG
Glutamate signaling may orchestrate oligodendrocyte precursor cell (OPC) development and myelin regeneration through the activation of glutamate receptors at OPC-neuron synapses. D-Aspartate is a D-amino acid exerting modulatory actions at glutamatergic synapses. Chronic administration of D-Aspartate has been proposed as therapeutic treatment in diseases related to myelin dysfunction and NMDA receptors hypofunction, including schizophrenia and cognitive deficits. Here, we show, by using an in vivo remyelination model, that administration of D-Aspartate during remyelination improved motor coordination, accelerated myelin recovery, and significantly increased the number of small-diameter myelinated axons. Chronically administered during demyelination, D-Aspartate also attenuated myelin loss and inflammation. Interestingly, D-Aspartate exposure stimulated OPC maturation and accelerated developmental myelination in organotypic cerebellar slices. D-Aspartate promoting effects on OPC maturation involved the activation of glutamate transporters, AMPA and NMDA receptors, and the Na+/Ca2+ exchanger NCX3. While blocking NMDA or NCX3 significantly prevented D-Aspartate-induced [Ca2+]i oscillations, blocking AMPA and glutamate transporters prevented both the initial and oscillatory [Ca2+]i response as well as D-Aspartate-induced inward currents in OPC Our findings reveal that D-Aspartate treatment may represent a novel strategy for promoting myelin recovery.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-Olig-2-Antikörper, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Neurofilament 200 in Kaninchen hergestellte Antikörper, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Myelin-Basisprotein-Antikörper, AS 82–87, culture supernatant, clone 12, Chemicon®
Sigma-Aldrich
Anti-Myelin-assoziierter Glykoprotein-Antikörper, Klon 513, clone 513, Chemicon®, from mouse