Merck
  • Characteristics of Waldenström Macroglobulinemia in Korean Patients According to Mutational Status of MYD88 and CXCR4: Analysis Using Ultra-Deep Sequencing.
Characteristics of Waldenström Macroglobulinemia in Korean Patients According to Mutational Status of MYD88 and CXCR4: Analysis Using Ultra-Deep Sequencing.
Clinical lymphoma, myeloma & leukemia (2019-06-22)
Dong Woo Shin, Sung-Min Kim, Jung-Ah Kim, Hee Sue Park, Sang Mee Hwang, Kyongok Im, Sungsik Kim, Jinhyun Kim, Sunghoon Kwon, Sung-Soo Yoon, Dong Soon Lee
ZUSAMMENFASSUNG
Little is known about the mutational frequency of myeloid differentiation factor 88 (MYD88) and C-X-C chemokine receptor type 4 (CXCR4) and the corresponding characteristics in Asian individuals afflicted with Waldenström macroglobulinemia (WM). We investigated the characteristics of WM according to mutational status of MYD88/CXCR4, and attempted to determine the lineage commitment among hematopoietic cells by MYD88L265P single-cell sequencing on bone marrow (BM) smear slides. CXCR4 mutations (muts) were detected using ultra-deep sequencing using target capture. Mutational burden of MYD88 was assessed using real-time polymerase chain reaction. Single-cell sequencing for MYD88 was performed on lymphocytes, plasmacytoid lymphocytes, plasma cells, and neutrophils using laser microdissection. Among 31 patients, the frequencies of MYD88/CXCR4 muts were as follows: MYD88 wild type (WT) CXCR4WT (6 patients, 19.4%), MYD88L265PCXCR4WT (19 patients, 61.4%), MYD88L265PCXCR4mut (6 patients, 19.4%; 1 frameshift and 5 nonsense muts). Immunoglobulin M levels of MYD88L265CXCR4WT patients were significantly higher than those of MYD88WTCXCR4WT patients (P = .024). Tumor burden in BM was highest in patients with MYD88L265PCXCR4mut (82.0%), followed by MYD88L265PCXCR4WT (52.8%) and MYD88WTCXCR4WT (14.2%) (P < .001). The quantity of MYD88-mutated DNA tended to correlate with tumor burden in BM (correlation coefficient 0.647; P = .009). MYD88L265P was detected in plasma cells, plasmacytoid lymphocytes, and lymphocytes but not neutrophils. The frequency of MYD88/CXCR4 muts in Korean and Caucasian patients with WM was similar, however 5 of the 6 CXCR4 muts were nonsense-a proportion higher than reported frequencies in Caucasian individuals. Ultra-deep sequencing was capable of detecting CXCR4 muts not detectable using Sanger sequencing, suggesting a possible replacement of the B-cell sorting.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteinase K aus Tritirachium album, buffered aqueous glycerol solution, Molecular Biology, ≥800 units/mL