Merck
The Structural Basis for the Binding of Repaglinide to the Pancreatic KATP Channel.
Cell reports (2019-05-09)
Dian Ding, Mengmeng Wang, Jing-Xiang Wu, Yunlu Kang, Lei Chen
ZUSAMMENFASSUNG
Repaglinide (RPG) is a short-acting insulin secretagogue widely prescribed for the treatment of type 2 diabetes. It boosts insulin secretion by inhibiting the pancreatic ATP-sensitive potassium channel (KATP). However, the mechanisms by which RPG binds to the KATP channel are poorly understood. Here, we describe two cryo-EM structures: the pancreatic KATP channel in complex with inhibitory RPG and adenosine-5'-(γ-thio)-triphosphate (ATPγS) at 3.3 Å and a medium-resolution structure of a RPG-bound mini SUR1 protein in which the N terminus of the inward-rectifying potassium channel 6.1 (Kir6.1) is fused to the ABC transporter module of the sulfonylurea receptor 1 (SUR1). These structures reveal the binding site of RPG in the SUR1 subunit. Furthermore, the high-resolution structure reveals the complex architecture of the ATP binding site, which is formed by both Kir6.2 and SUR1 subunits, and the domain-domain interaction interfaces.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Adenosin-5′-[γ-thio]triphosphat Tetralithiumsalz, ≥75% (HPLC), powder
Sigma-Aldrich
Diosgenin, ≥93%
Sigma-Aldrich
Indole-3-butyric acid potassium salt, suitable for plant cell culture, BioReagent
Sigma-Aldrich
Repaglinid, ≥98% (HPLC), solid