Merck
  • CAMSAP2 Is a Microtubule Minus-End Targeting Protein That Regulates BTB Dynamics Through Cytoskeletal Organization.
CAMSAP2 Is a Microtubule Minus-End Targeting Protein That Regulates BTB Dynamics Through Cytoskeletal Organization.
Endocrinology (2019-04-18)
Bai-Ping Mao, Linxi Li, Renshan Ge, Chao Li, Chris K C Wong, Bruno Silvestrini, Qingquan Lian, C Yan Cheng
ZUSAMMENFASSUNG
During spermatogenesis, microtubule (MT) cytoskeleton in Sertoli cells confers blood-testis barrier (BTB) function, but the regulators and mechanisms that modulate MT dynamics remain unexplored. In this study, we examined the role of calmodulin-regulated spectrin-associated protein (CAMSAP)2 (a member of the CAMSAP/Patronin protein family), and a minus-end targeting protein (-TIP) that binds to the minus-end (i.e., slow-growing end) of polarized MTs involved in determining MT length, in Sertoli cell function. CAMSAP2 was found to localize at discrete sites across the Sertoli cell cytosol, different from end-binding protein 1 (a microtubule plus-end tracking protein that binds to the plus-end of MTs), and colocalized with MTs. CAMSAP2 displayed a stage-specific expression pattern, appearing as tracklike structures across the seminiferous epithelium in adult rat testes that lay perpendicular to the basement membrane. CAMSAP2 knockdown by RNA interference was found to promote Sertoli cell tight junction (TJ) barrier function, illustrating its role in inducing TJ remodeling under physiological conditions. To further examine the regulatory role of CAMSAP2 in BTB dynamics, we used a perfluorooctanesulfonate (PFOS)-induced Sertoli cell injury model for investigations. CAMSAP2 knockdown blocked PFOS-induced Sertoli cell injury by promoting proper distribution of BTB-associated proteins at the cell-cell interface. This effect was mediated by the ability of CAMSAP2 knockdown to block PFOS-induced disruptive organization of MTs, but also F-actin, across cell cytosol through changes in cellular distribution/localization of MT- and actin-regulatory proteins. In summary, CAMSAP2 is a regulator of MT and actin dynamics in Sertoli cells to support BTB dynamics and spermatogenesis.