Merck
  • JMJD6 regulates histone H2A.X phosphorylation and promotes autophagy in triple-negative breast cancer cells via a novel tyrosine kinase activity.
JMJD6 regulates histone H2A.X phosphorylation and promotes autophagy in triple-negative breast cancer cells via a novel tyrosine kinase activity.
Oncogene (2018-09-07)
Yan Liu, Yue-Hong Long, Shu-Qing Wang, Yuan-Yue Zhang, Yu-Feng Li, Jiang-Sheng Mi, Cheng-Hua Yu, De-Yan Li, Jing-Hua Zhang, Xiao-Jun Zhang
ZUSAMMENFASSUNG
Overexpression of Jumonji domain-containing 6 (JMJD6) has been reported to be associated with more aggressive breast cancer characteristics. However, the precise role of JMJD6 in breast cancer development remains unclear. Here, we demonstrate that JMJD6 has intrinsic tyrosine kinase activity and can utilize ATP and GTP as phosphate donors to phosphorylate Y39 of histone H2A.X (H2A.XY39ph). High JMJD6 levels promoted autophagy in triple negative breast cancer (TNBC) cells by regulating the expression of autophagy-related genes. The JMJD6-H2A.XY39ph axis promoted TNBC cell growth via the autophagy pathway. We show that combined inhibition of JMJD6 kinase activity and autophagy efficiently decreases TNBC growth. Together, these findings suggest an effective strategy for TNBC treatment.