Merck
Physiological and Metabolomic Analysis of Cold Plasma Treated Fresh-Cut Strawberries.
Journal of agricultural and food chemistry (2019-03-19)
Meilin Li, Xiaoan Li, Cong Han, Nana Ji, Peng Jin, Yonghua Zheng
ZUSAMMENFASSUNG
Cold plasma technology offers new opportunities to the decontamination and preservation of fruits and vegetables. In the present research, strawberries were cut into four wedges and then treated with dielectric barrier discharge plasma at 45 kV for 1 min and stored for 1 week (4 °C). Metabolomic analysis suggested that plasma treatment improved the biosynthesis of the metabolites in the "flavones and flavonol biosynthesis" pathway and "biosynthesis of phenylpropanoids" pathway in fresh-cut strawberries. Physiological assay demonstrated that plasma treatment maintained the texture properties and inhibited microbial growth of fresh-cut strawberries. In addition, plasma treatment also promoted the accumulation of total phenolics, total flavonoid, and anthocyanin by enhancing the critical enzyme activities and activating related gene expression in phenylpropanoid as well as reactive oxygen species metabolism, which contributed greatly to the enhancement of antioxidant capacity of strawberry wedges. Our investigation provided a new perspective of the effect of plasma treatment on the safety and quality of strawberry wedges and suggested that cold plasma treatment holds promise as an emerging processing technology for improving the quality and antioxidant activity of postharvest fruits and vegetables.