Merck
  • Serological and histological evaluation of species-specific immunocontraceptive vaccine antigens based on zona pellucida 3 in the small Indian mongoose (Herpestes auropunctatus).
Serological and histological evaluation of species-specific immunocontraceptive vaccine antigens based on zona pellucida 3 in the small Indian mongoose (Herpestes auropunctatus).
The Journal of veterinary medical science (2018-12-28)
Naotoshi Kuninaga, Makoto Asano, Ryota Matsuyama, Takahiro Minemoto, Takayuki Mori, Masatsugu Suzuki
ZUSAMMENFASSUNG
The small Indian mongoose (Herpestes auropunctatus) was introduced to Japanese islands and has impacted on the island's biodiversity. Population control has been attempted through capturing but its efficiency has rapidly declined. Therefore, new additional control methods are required. Our focus has been on the immunocontraceptive vaccines, which act in an especially species-specific manner. The amino-acid sequence of the mongoose ovum zona pellucida protein 3 (ZP3) was decoded and two types of synthetic peptides (A and B) were produced. In this study, these peptides were administered to mongooses (each n=3) and the sera were collected to verify immunogenicity using ELISA and IHC. Treated mongoose sera showed an increasing of antibody titer according to immunizations and the antigen-antibody reactions against the endogenous mongoose ZP. In addition, IHC revealed that immune sera absorbed with each peptide showed a marked reduction in reactivity, which indicated the specificity of induced antibodies. These reactions were marked in peptide A treated mongoose sera, and the antibody titer of one of them lasted for at least 21 weeks. These results indicated that peptide A was a potential antigen, inducing autoantibody generation. Moreover, immunized rabbit antibodies recognized mongoose ZP species-specifically. However, the induction of robust immune memory was not observed. Also, the actual sterility effects of peptides remain unknown, it should be verified as a next step. In any case, this study verified synthetic peptides we developed are useful as the antigen candidates for immunocontraception of mongooses.