Merck
  • PD-L1 Expression of Lung Cancer Cells, Unlike Infiltrating Immune Cells, Is Stable and Unaffected by Therapy During Brain Metastasis.
PD-L1 Expression of Lung Cancer Cells, Unlike Infiltrating Immune Cells, Is Stable and Unaffected by Therapy During Brain Metastasis.
Clinical lung cancer (2019-06-11)
Vanda Téglási, Orsolya Pipek, Rita Lózsa, Kinga Berta, Dávid Szüts, Tünde Harkó, Pál Vadász, Lívia Rojkó, Balázs Döme, Attila G Bagó, József Tímár, Judit Moldvay, Zoltán Szállási, Lilla Reiniger
ZUSAMMENFASSUNG
Approximately 50% of brain metastases originate from non-small-cell lung cancer. The median survival of patients with brain metastases is 1 month without treatment. Novel immunotherapeutic strategies, such as those targeting the programmed death ligand 1 (PD-L1)/programmed cell death 1 (PD-1) axis, are promising in patients with advanced systemic disease but are often preferentially administered to patients with tumors showing PD-L1 positivity. Surgically resected paired primary lung adenocarcinoma and brain metastasis samples of 61 patients were analyzed. We compared the paired samples regarding the amount of peritumoral and stromal mononuclear infiltration, PD-L1 expression of tumor and immune cells, and PD-1 expression of immune cells. We investigated the effect of radiotherapy, chemotherapy, and steroid therapy on PD-L1 expression in brain metastases. There was significant positive correlation regarding the PD-L1 expression of tumor cells between the paired primary lung adenocarcinoma and brain metastatic samples with the use of different cutoff levels (1%, 5%, 50%). We found no impact of chemotherapy or steroid therapy on the changes of PD-L1 expression of tumor cells between the 2 sites. There is no or only limited concordance of the proportion of PD-1- or PD-L1-positive tumor-associated immune cells between the paired tumor samples, which suggests that brain metastases develop their own immune environment. We observed a strong correlation of PD-L1 positive tumor cells between primary lung adenocarcinoma cases and their corresponding brain metastases, which is not significantly influenced by chemotherapy or steroid therapy.