Merck
High molecular weight amyloid β1-42 oligomers induce neurotoxicity via plasma membrane damage.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2019-05-16)
Taro Yasumoto, Yusaku Takamura, Mayumi Tsuji, Takahiro Watanabe-Nakayama, Keiko Imamura, Haruhisa Inoue, Shiro Nakamura, Tomio Inoue, Atsushi Kimura, Satoshi Yano, Hisao Nishijo, Yuji Kiuchi, David B Teplow, Kenjiro Ono
ZUSAMMENFASSUNG
Amyloid β-protein (Aβ) molecules tend to aggregate and subsequently form low MW (LMW) oligomers, high MW (HMW) aggregates such as protofibrils, and ultimately fibrils. These Aβ species can generally form amyloid plaques implicated in the neurodegeneration of Alzheimer disease (AD), but therapies designed to reduce plaque load have not demonstrated clinical efficacy. Recent evidence implicates amyloid oligomers in AD neuropathology, but the precise mechanisms are uncertain. We examined the mechanisms of neuronal dysfunction from HMW-Aβ1-42 exposure by measuring membrane integrity, reactive oxygen species (ROS) generation, membrane lipid peroxidation, membrane fluidity, intracellular calcium regulation, passive membrane electrophysiological properties, and long-term potentiation (LTP). HMW-Aβ1-42 disturbed membrane integrity by inducing ROS generation and lipid peroxidation, resulting in decreased membrane fluidity, intracellular calcium dysregulation, depolarization, and impaired LTP. The damaging effects of HMW-Aβ1-42 were significantly greater than those of LMW-Aβ1-42. Therapeutic reduction of HMW-Aβ1-42 may prevent AD progression by ameliorating direct neuronal membrane damage.-Yasumoto, T., Takamura, Y., Tsuji, M., Watanabe-Nakayama, T., Imamura, K., Inoue, H., Nakamura, S., Inoue, T., Kimura, A., Yano, S., Nishijo, H., Kiuchi, Y., Teplow, D. B., Ono, K. High molecular weight amyloid β1-42 oligomers induce neurotoxicity via plasma membrane damage.