Merck
  • Prostaglandin E2 inhibits profibrotic function of human pulmonary fibroblasts by disrupting Ca2+ signaling.
Prostaglandin E2 inhibits profibrotic function of human pulmonary fibroblasts by disrupting Ca2+ signaling.
American journal of physiology. Lung cellular and molecular physiology (2019-02-14)
Subhendu Mukherjee, Wei Sheng, Alexander Michkov, Krishna Sriarm, Rui Sun, Anna Dvorkin-Gheva, Paul A Insel, Luke J Janssen
ZUSAMMENFASSUNG
We have shown that calcium (Ca2+) oscillations in human pulmonary fibroblasts (HPFs) contribute to profibrotic effects of transforming growth factor-β (TGF-β) and that disruption of these oscillations blunts features of pulmonary fibrosis. Prostaglandin E2 (PGE2) exerts antifibrotic effects in the lung, but the mechanisms for this action are not well defined. We thus sought to explore interactions between PGE2 and the profibrotic agent TGF-β in pulmonary fibroblasts (PFs) isolated from patients with or without idiopathic pulmonary fibrosis (IPF). PGE2 inhibited TGF-β-promoted [Ca2+] oscillations and prevented the activation of Akt and Ca2+/calmodulin-dependent protein kinase-II (CaMK-II) but did not prevent activation of Smad-2 or ERK. PGE2 also eliminated TGF-β-stimulated expression of collagen A1, fibronectin, and α-smooth muscle actin and reduced stress fiber formation in the HPFs. RNA sequencing revealed that HPFs preferentially express EP2 receptors relative to other prostanoid receptor subtypes: EP2 expression is ~10-fold higher than that of EP4 receptors; EP1 and EP3 receptors are barely detectable; and EP2-receptor expression is ~3.5-fold lower in PFs from IPF patients than in normal HPFs. The inhibitory effects of PGE2 on synthetic function and stress fiber formation were blocked by selective EP2 or EP4 antagonists and mimicked by selective EP2 or EP4 agonists, the phosphodiesterase inhibitor isobutylmethylxanthine and forskolin, all of which elevate cellular cAMP concentrations. We conclude that PGE2, likely predominantly via EP2 receptors, interferes with Ca2+ signaling, CaMK-II activation, and Akt activation in IPF-HPFs and HPFs treated with TGF-β. Moreover, a decreased expression of EP2 receptors in pulmonary fibroblasts from IPF patients may contribute to the pathophysiology of this disease.