Merck
Direct Induction of the Three Pre-implantation Blastocyst Cell Types from Fibroblasts.
Cell stem cell (2019-04-30)
Hana Benchetrit, Mohammad Jaber, Valery Zayat, Shulamit Sebban, Avital Pushett, Kirill Makedonski, Zvi Zakheim, Ahmed Radwan, Noam Maoz, Rachel Lasry, Noa Renous, Michal Inbar, Oren Ram, Tommy Kaplan, Yosef Buganim
ZUSAMMENFASSUNG
Following fertilization, totipotent cells undergo asymmetric cell divisions, resulting in three distinct cell types in the late pre-implantation blastocyst: epiblast (Epi), primitive endoderm (PrE), and trophectoderm (TE). Here, we aim to understand whether these three cell types can be induced from fibroblasts by one combination of transcription factors. By utilizing a sophisticated fluorescent knockin reporter system, we identified a combination of five transcription factors, Gata3, Eomes, Tfap2c, Myc, and Esrrb, that can reprogram fibroblasts into induced pluripotent stem cells (iPSCs), induced trophoblast stem cells (iTSCs), and induced extraembryonic endoderm stem cells (iXENs), concomitantly. In-depth transcriptomic, chromatin, and epigenetic analyses provide insights into the molecular mechanisms that underlie the reprogramming process toward the three cell types. Mechanistically, we show that the interplay between Esrrb and Eomes during the reprogramming process determines cell fate, where high levels of Esrrb induce a XEN-like state that drives pluripotency and high levels of Eomes drive trophectodermal fate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti-Dimethylhiston-H3-(Lys4)-Antikörper, Upstate®, from rabbit