Merck
  • lncRNA-ES3/miR-34c-5p/BMF axis is involved in regulating high-glucose-induced calcification/senescence of VSMCs.
lncRNA-ES3/miR-34c-5p/BMF axis is involved in regulating high-glucose-induced calcification/senescence of VSMCs.
Aging (2019-01-18)
Xiao Lin, Jun-Kun Zhan, Jia-Yu Zhong, Yan-Jiao Wang, Yi Wang, Shuang Li, Jie-Yu He, Pan Tan, Yi-Yin Chen, Xue-Bin Liu, Xing-Jun Cui, You-Shuo Liu
ZUSAMMENFASSUNG
Vascular calcification/aging is common in diabetes and is associated with increased morbidity and mortality of patients. MiR-34c-5p, not miR-34c-3p, was suppressed significantly in calcification/senescence of human aorta vascular smooth muscle cells (HA-VSMCs) induced by high glucose, which was proven by the formation of mineralized nodules and staining of senescence associated-β-galactosidase staining (SA β-gal) positive cells. Overexpression of miR-34c-5p alleviated calcification/senescence of HA-VSMCs, whereas inhibition of miR-34c-5p received the opposite results. Bcl-2 modifying factor (BMF) was a functional target of miR-34c-5p and it was involved in the process of calcification/senescence of HA-VSMCs. Besides, lncRNA-ES3 acted as a competing endogenous RNAs (ceRNA) of miR-34c-5p to enhance BMF expression. Further, lncRNA-ES3 inhibited miR-34c-5p expression by direct interaction and its knockdown suppressed the calcification/senescence of HA-VSMCs. Our results showed for the first time that the calcification/senescence of VSMCs was regulated by lncRNA-ES3 /miR-34c-5p/BMF axis.