Merck
  • In Vitro Infant Faecal Fermentation of Low Viscosity Barley β-Glucan and Its Acid Hydrolyzed Derivatives: Evaluation of Their Potential as Novel Prebiotics.
In Vitro Infant Faecal Fermentation of Low Viscosity Barley β-Glucan and Its Acid Hydrolyzed Derivatives: Evaluation of Their Potential as Novel Prebiotics.
Molecules (Basel, Switzerland) (2019-03-01)
Ka-Lung Lam, Kin-Chun Ko, Xiaojie Li, Xinxin Ke, Wai-Yin Cheng, Tianfeng Chen, Lijun You, Hoi-Shan Kwan, Peter Chi-Keung Cheung
ZUSAMMENFASSUNG
Barley contains high level of β-1,3-1,4-glucans (BBGs) which can be fermented by microbes and are a potential prebiotic. In the present study, native BBG with low viscosity and a MW of 319 kDa was depolymerized by acid hydrolysis to produce a series of four structurally characterized fragments with MWs ranging from 6⁻104 kDa. In vitro fermentation of these BBG samples by infant faecal microbiome was evaluated using a validated deep-well plate protocol as parallel miniature bioreactors. Microbial taxa were identified using 16S amplicon sequencing after 40 h of anaerobic fermentation. Bioinformatics analysis including diversity indexes, predicted metagenomic KEGG functions and predicted phenotypes were performed on the sequenced data. Short chain fatty acids and dissolved ammonia were quantified and the SCFAs/NH₃ ratio was used to evaluate the eubiosis/dysbiosis potential. Correlation analysis showed that most of the parameters investigated showed a parabolic function instead of a monotonous function with the BBG samples having different MWs. Among the five BBGs, it was concluded that BBG with an intermediate MW of 28 kDa is the most promising candidate to be developed as a novel prebiotic.