Merck
  • Nanocrystalline solid dispersions (NSD) of hesperetin (HRN) for prevention of 7, 12-dimethylbenz[a]anthracene (DMBA)-induced breast cancer in Sprague-Dawley (SD) rats.
Nanocrystalline solid dispersions (NSD) of hesperetin (HRN) for prevention of 7, 12-dimethylbenz[a]anthracene (DMBA)-induced breast cancer in Sprague-Dawley (SD) rats.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2018-12-16)
Sneha Sheokand, Umashanker Navik, Arvind K Bansal
ZUSAMMENFASSUNG
Breast cancer is the most commonly diagnosed cancer and the leading cause of cancer death in females as per the global cancer project (GLOBOCAN 2018) estimates of breast cancer incidence and mortality produced by the International Agency for Research on Cancer (IARC). In 2018, there will be 2,088,849 new cases of breast cancer and 626,679 cases of deaths due to breast cancer in 5 regions (Americas, Africa, Europe, Asia and Oceania) at 20 sites of the world. Epidemiologic studies on diet and cancer have guided the search for some nutraceuticals acting as anti-cancer agents. Hesperetin (HRN), the aglycone of hesperidin, a glycoside found in citrus fruits, has been reported to have anti-cancer effects by apoptosis induction and inhibition of cell proliferation in cancer cells. However, poor solubility of HRN has limited its absorption while deviating from its therapeutic benefit. The present study aimed to develop nanocrystalline solid dispersions (NSD) of HRN and evaluating the oral bioavailability in rats. The study also evaluated the efficacy of NSDs against the carcinogenic activity of DMBA in female rats. NSDs were optimized using design of experiments (DoE) and multivariate analysis (MVA) tools. The optimized NSD formulation showed an average particle size (Zavg) of 558.2 ± 68.1 nm and ~70% release in 30 min. The in vivo pharmacokinetic study also construed remarkable improvement (3.3 and 2.1-fold increase in Cmax and AUC0-∞) in rate and extent of absorption and 4-fold reduction in Tmax by the optimized NSD formulation. In vivo chemoprevention study construed superior efficacy of the NSD formulation by reducing the tumor burden, delaying the onset of tumors and reducing the tumor weight and volume in DMBA-induced breast cancer rats. In conclusion, we present a simple NSD formulation of HRN with enhanced bioavailability and superior chemopreventive efficacy.