Merck
  • An untargeted metabolomic strategy based on liquid chromatography-mass spectrometry to study high glucose-induced changes in HK-2 cells.
An untargeted metabolomic strategy based on liquid chromatography-mass spectrometry to study high glucose-induced changes in HK-2 cells.
Journal of chromatography. A (2019-03-18)
Samuel Bernardo-Bermejo, Elena Sánchez-López, María Castro-Puyana, Selma Benito, Francisco Javier Lucio-Cazaña, María Luisa Marina
ZUSAMMENFASSUNG
Diabetes mellitus is a major health concern nowadays. It is estimated that 40% of diabetics are affected by diabetic nephropathy, one of the complications derived from high glucose blood levels which can lead to chronic loss of kidney function. It is now clear that the renal proximal tubule plays a critical role in the progression of diabetic nephropathy but research focused on studying the molecular mechanisms involved is still needed. The aim of this work was to develop a liquid chromatography-mass spectrometry platform to carry out, for the first time, the untargeted metabolomic analysis of high glucose-induced changes in cultured human proximal tubular HK-2 cells. In order to find the metabolites which were affected by high glucose and to expand the metabolite coverage, intra- and extracellular fluid from HK-2 cells exposed to high glucose (25 mM), normal glucose (5.5 mM) or osmotic control (5.5 mM glucose +19.5 mM mannitol) were analyzed by two complementary chromatographic modes: hydrophilic interaction and reversed-phase liquid chromatography. Non-supervised principal components analysis showed a good separation among the three groups of samples. Statistically significant variables were chosen for further metabolite identification. Different metabolic pathways were affected mainly those derived from amino acidic, polyol, and nitrogenous bases metabolism.