Merck
CBX3 predicts an unfavorable prognosis and promotes tumorigenesis in osteosarcoma.
Molecular medicine reports (2019-04-04)
Chao Ma, Xing-Guo Nie, Yan-Li Wang, Xiang-Hua Liu, Xue Liang, Qing-Lan Zhou, Da-Peng Wu
ZUSAMMENFASSUNG
CBX3, namely chromobox protein homolog 3, a member of the heterochomatin protein 1 (HP1) family, has been shown to be associated with the tumorigenesis of various types of cancer. The aim of the present study was to assess the biological role and the clinicopathological importance of CBX3 in osteosarcoma. The Oncomine database was utilized to determine the CBX3 expression in sarcoma patients. A retrospective cohort study was conducted to evaluate the prognostic value of CBX3 expression. In addition, correlations between the clinicopathological features of the osteosarcoma patients and CBX3 expression were assessed and involved recurrence, distant metastasis, lymph node metastasis, response to chemotherapy, pathological differentiation, clinical stage, anatomic location, tumor size and age. To investigate the function of CBX3 in osteosarcoma, a small interfering RNA for CBX3 was designed and this was used for the transfection of osteosarcoma MG63 cells. Then, the effects of CBX3 on proliferation, cell cycle distribution and apoptosis of osteosarcoma cells were investigated via CCK‑8 assay and cell cycle assay and cell apoptosis analysis, respectively. Based on our findings, upregulation of CBX3 expression was noted both in osteosarcoma and also other sarcoma types, which included pleomorphic liposarcoma, myxofibrosarcoma, myxoid/round cell liposarcoma and dedifferentiated liposarcoma. In addition, based on the retrospective cohort study, CBX3 expression was associated with the disease‑free survival (DFS) and overall survival (OS) of the osteosarcoma patients and a large tumor size, high distant metastasis rate and high clinical stage rate. In addition, the proliferation ability was blocked by the knockdown of CBX3 through the application of CBX3 siRNA, and CBX3 knockdown also led to increased apoptosis and cell cycle arrest at G0 and G1 phases in osteosarcoma cells. CBX3 is highly expressed in human osteosarcoma tissues. Meanwhile, high CBX3 is a predictor of the poor prognosis of osteosarcoma patients. To conclude, the growth of osteosarcoma can be promoted by CBX3, which may be used as an independent potential prognostic biomarker for patients suffering from osteosarcoma.