Merck
  • The laminin binding α3 and α6 integrins cooperate to promote epithelial cell adhesion and growth.
The laminin binding α3 and α6 integrins cooperate to promote epithelial cell adhesion and growth.
Matrix biology : journal of the International Society for Matrix Biology (2018-09-09)
Eugenia M Yazlovitskaya, Olga M Viquez, Tianxiang Tu, Adele De Arcangelis, Elisabeth Georges-Labouesse, Arnoud Sonnenberg, Ambra Pozzi, Roy Zent
ZUSAMMENFASSUNG
Integrins, the major receptors for cell-extracellular matrix (ECM) interactions, regulate multiple cell biological processes including adhesion, migration, proliferation and growth factor-dependent signaling. The principal laminin (LM) binding integrins α3β1, α6β1 and α6β4 are usually co-expressed in cells and bind to multiple laminins with different affinities making it difficult to define their specific function. In this study, we generated kidney epithelial collecting duct (CD) cells that lack both the α3 and α6 integrin subunits. This deletion impaired cell adhesion and migration to LM-332 and LM-511 more than deleting α3 or α6 alone. Cell adhesion mediated by both α3β1 and α6 integrins was PI3K independent, but required K63-linked polyubiquitination of Akt by the ubiquitin-modifying enzyme TRAF6. Moreover, we provide evidence that glial-derived neurotrophic factor (GDNF) and fibroblast growth factor 10 (FGF10)- mediated cell signaling, spreading and proliferation were severely compromised in double integrin α3/α6- but not single α3- or α6-null CD cells. Interestingly, these growth factor-dependent cell functions required both PI3K- and TRAF6-dependent Akt activation. These data suggest that expression of the integrin α3 or α6 subunit is sufficient to mediate GDNF- and FGF10-dependent spreading, proliferation and signaling on LM-511. Thus, our study shows that α3 and α6 containing integrins promote distinct functions and signaling by CD cells on laminin substrata.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-Aktin in Maus hergestellte Antikörper, clone AC-40, ascites fluid
Sigma-Aldrich
Anti-Integrin-α3-Antikörper, serum, Chemicon®