Merck
  • Mitochondrial Hyperactivation and Enhanced ROS Production are Involved in Toxicity Induced by Oncogenic Kinases Over-Signaling.
Mitochondrial Hyperactivation and Enhanced ROS Production are Involved in Toxicity Induced by Oncogenic Kinases Over-Signaling.
Cancers (2018-12-14)
Monica Ceccon, Mario Mauri, Luca Massimino, Giovanni Giudici, Rocco Piazza, Carlo Gambacorti-Passerini, Luca Mologni
ZUSAMMENFASSUNG
Targeted therapy is an effective, rational, and safe approach to solid and hematological tumors treatment. Unfortunately, a significant fraction of patients treated with tyrosine kinase inhibitors (TKI) relapses mainly because of gene amplification, mutations, or other bypass mechanisms. Recently a growing number of papers showed how, in some cases, resistance due to oncogene overexpression may be associated with drug addiction: cells able to proliferate in the presence of high TKI doses become also TKI dependent, undergoing cellular stress, and apoptosis/death upon drug withdrawal. Notably, if a sub-cellular population survives TKI discontinuation it is also partially re-sensitized to the same drug. Thus, it is possible that a subset of patients relapsing upon TKI treatment may benefit from a discontinuous therapeutic schedule. We focused on two different hematologic malignancies, chronic myeloid leukemia (CML) and anaplastic large cell lymphoma (ALCL), both successfully treatable with TKIs. The two models utilized (LAMA and SUP-M2) differed in having oncogene overexpression as the sole cause of drug resistance (CML), or additionally carrying kinase domain mutations (ALCL). In both cases drug withdrawal caused a sudden overload of oncogenic signal, enhanced mitochondria activity, induced the release of a high amount of reactive oxygen species (ROS), and caused genotoxic stress and massive cell death. In LAMA cells (CML) we could rescue the cells from death by partially blocking downstream oncogenic signaling or lowering ROS detrimental effect by adding reduced glutathione.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, purified immunoglobulin, buffered aqueous solution