Merck
Teleost Gasdermin E Is Cleaved by Caspase 1, 3, and 7 and Induces Pyroptosis.
Journal of immunology (Baltimore, Md. : 1950) (2019-08-09)
Shuai Jiang, Hanjie Gu, Yan Zhao, Li Sun
ZUSAMMENFASSUNG
Pyroptosis is a newly defined gasdermin (GSDM)-dependent inflammatory type of programmed cell death. Different from mammals, which have a panel of pyroptotic GSDM members (e.g., GSDMA-E), teleosts possess only GSDME. The pyroptotic activity and regulation mechanism of teleost GSDME remain to be elucidated. In this work, we investigated the activity of the teleost Cynoglossus semilaevis (tongue sole) GSDME (CsGSDME) in association with different caspases (CASPs). We found that CsGSDME exerted pyroptotic and bactericidal activities through its N-terminal domain. Unlike human GSDME, which is exclusively cleaved by CASP3, CsGSDME was cleaved by C. semilaevis CASP (CsCASP) 1 with high efficiency and by CsCASP3 and 7 with comparatively low efficiencies, and all cleavages occurred at the 243FEVD246 site in the interdomain linker region of CsGSDME. Mutation of Phe243 to Asp/Ala and Asp246 to Ala in 243FEVD246 altered the cleavage preference of CsCASP1, 3, and 7. Treatment with loss-of-function CsCASP mutants or inhibition of CsCASP activity resulted in failure of CsGSDME cleavage. CsCASP1-cleaved CsGSDME induced pyroptosis, whereas CsCASP3/7-cleaved CsGSDME and F243 mutants induced switching of cell death from apoptosis to pyroptosis. Analysis of 54 teleost GSDME sequences revealed a conserved tetrapeptide motif that fits well to the inherent cleavage site of CASP1. Taken together, the results of our study demonstrate a hitherto, to our knowledge, unrecognized GSDME cleavage mode in teleosts that is clearly different from that in mammals, thus providing an important insight into the activation mechanism of CASP-mediated, GSDM-executed pyroptosis in teleosts.