Merck
  • Novel Central Nervous System (CNS)-Targeting Protease Inhibitors for Drug-Resistant HIV Infection and HIV-Associated CNS Complications.
Novel Central Nervous System (CNS)-Targeting Protease Inhibitors for Drug-Resistant HIV Infection and HIV-Associated CNS Complications.
Antimicrobial agents and chemotherapy (2019-05-08)
Masayuki Amano, Pedro Miguel Salcedo-Gómez, Ravikiran S Yedidi, Rui Zhao, Hironori Hayashi, Kazuya Hasegawa, Tomofumi Nakamura, Cuthbert D Martyr, Arun K Ghosh, Hiroaki Mitsuya
ZUSAMMENFASSUNG
There is currently no specific therapeutics for the HIV-1-related central nervous system (CNS) complications. Here we report that three newly designed CNS-targeting HIV-1 protease inhibitors (PIs), GRL-083-13, GRL-084-13, and GRL-087-13, which contain a P1-3,5-bis-fluorophenyl or P1-para-monofluorophenyl ring, and P2-bis-tetrahydrofuran (bis-THF) or P2-tetrahydropyrano-tetrahydrofuran (Tp-THF), with a sulfonamide isostere, are highly active against wild-type HIV-1 strains and primary clinical isolates (50% effective concentration [EC50], 0.0002 to ∼0.003 μM), with minimal cytotoxicity. These CNS-targeting PIs efficiently suppressed the replication of HIV-1 variants (EC50, 0.002 to ∼0.047 μM) that had been selected to propagate at high concentrations of conventional HIV-1 PIs. Such CNS-targeting PIs maintained their antiviral activity against HIV-2ROD as well as multidrug-resistant clinical HIV-1 variants isolated from AIDS patients who no longer responded to existing antiviral regimens after long-term therapy. Long-term drug selection experiments revealed that the emergence of resistant-HIV-1 against these CNS-targeting PIs was substantially delayed. In addition, the CNS-targeting PIs showed the most favorable CNS penetration properties among the tested compounds, including various FDA-approved anti-HIV-1 drugs, as assessed with the in vitro blood-brain barrier reconstruction system. Crystallographic analysis demonstrated that the bicyclic rings at the P2 moiety of the CNS-targeting PIs form strong hydrogen-bond interactions with HIV-1 protease (PR) active site. Moreover, both the P1-3,5-bis-fluorophenyl and P1-para-monofluorophenyl rings sustain greater van der Waals contacts with PR than in the case of darunavir (DRV). The data suggest that the present CNS-targeting PIs have desirable features for treating patients infected with wild-type and/or multidrug-resistant HIV-1 strains and might serve as promising preventive and/or therapeutic candidates for HIV-1-associated neurocognitive disorders (HAND) and other CNS complications.