Merck
  • Risk factors for early and late adenoma recurrence after advanced colorectal endoscopic resection at an expert Western center.
Risk factors for early and late adenoma recurrence after advanced colorectal endoscopic resection at an expert Western center.
Gastrointestinal endoscopy (2019-03-03)
Andrew Emmanuel, Christo Lapa, Anil Ghosh, Shraddha Gulati, Margaret Burt, Bu'Hussain Hayee, Amyn Haji
ZUSAMMENFASSUNG
Few large Western series examine risk factors for recurrence after endoscopic resection (ER) of large (≥20 mm) colorectal laterally spreading tumors. Recurrence beyond initial surveillance is seldom reported, and differences between residual/recurrent adenoma and late recurrence are not scrutinized. We report the incidence of recurrence at successive surveillance intervals, identify risk factors for recurrent/residual adenoma and late recurrence, and describe the outcomes of ER of recurrent adenomas. Recurrence was calculated for successive surveillance periods after colorectal ER. Multiple logistic regression was used to identify independent risk factors for recurrent/residual adenoma and late recurrence (≥12 months). Six hundred twenty colorectal ERs were performed, and 456 eligible patients (98%) had completed 3- to 6-month surveillance. Residual/recurrent adenoma (3-6 months) was detected in 8.3%, at 12 months in 6.1%, between 24 and 36 months in 6.4%, and after 36 months in 13.5%. Independent risk factors for residual/recurrent adenoma were piecemeal resection (odds ratio [OR], 13.0; P = .01), adjunctive argon plasma coagulation (OR, 2.4; P = .01), and lesion occupying ≥75% of the luminal circumference (OR, 5.6; P < .001) and for late recurrence were lesion size >60 mm (OR, 6.3; P < .001) and piecemeal resection (OR, 4.4; P = .04). Of 66 patients with recurrence, 5 required surgery, 8 left the treatment pathway, 20 are still receiving ER or surveillance, and 33 had ER with normal subsequent surveillance. Recurrence occurs at successive periods of surveillance after ER even beyond 3 years. Aside from piecemeal resection, risk factors for residual/recurrent adenoma and late recurrence are different. Recurrence can be challenging to treat, but surgery is rarely required.