Merck
  • MicroRNA-145 performs as a tumor suppressor in human esophageal squamous cell carcinoma by targeting phospholipase C epsilon 1.
MicroRNA-145 performs as a tumor suppressor in human esophageal squamous cell carcinoma by targeting phospholipase C epsilon 1.
Journal of cellular biochemistry (2019-01-18)
Chun Tang, Jin-Yuan He, Chao Yu, Pei-Jie Wang, Shao-Hong Huang, Hong-Jie Zheng, Dong-Qing Yan, Jun-Hang Zhang, Yun Li
ZUSAMMENFASSUNG
Esophageal squamous cell carcinoma (ESCC) is the leading pathologic type in China. miR-145 has been reported to be downregulated in multiple tumors. This study was aimed to investigate the role of miR-145 in ESCC. miR-145 expression was investigated in 65 ESCC samples as well as four ESCC cell lines by quantitative real-time polymerase chain reaction (qRT-PCR). Targetscan 6.2 website (http://www.targetscan.org/) was used to predict the targets of miR-145. Expression of phospholipase C epsilon 1 (PLCE1) messenger RNA and protein was detected by qRT-PCR or Western blot. MTT and wound healing assay were conducted to explore the effects of miR-145 on the proliferation and migration of ESCC cell lines, respectively. miR-145 was significantly decreased in ESCC tissues. An inverse correlation between miR-145 and invasion depth and TNM stage were observed. PLCE1 was a direct target of miR-145, and the expression of PLCE1 was inversely correlated with miR-145 expression in ESCC tissues. In addition, overexpression of miR-145 suppressed cell proliferation and migration in ESCC cells. The enforced expression of PLCE1 partially reversed the suppressive effect of miR-145. These results prove that miR-145 may perform as a tumor suppressor in ESCC by targeting PLCE1.