Merck
  • Effects of MCRS1 on proliferation, migration, invasion, and epithelial mesenchymal transition of gastric cancer cells by interacting with Pkmyt1 protein kinase.
Effects of MCRS1 on proliferation, migration, invasion, and epithelial mesenchymal transition of gastric cancer cells by interacting with Pkmyt1 protein kinase.
Cellular signalling (2019-04-07)
Xin-Meng Wang, Qi-Yang Li, Li-Li Ren, Yi-Meng Liu, Tian-Shi Wang, Tian-Chi Mu, Shuai Fu, Chao Liu, Jian-Ying Xiao
ZUSAMMENFASSUNG
Microspherule protein 1(MCRS1) is known to be an oncogene in several tumors. However, recent studies have shown that MCRS1 inhibits lymphatic metastasis in gastric cancer (GC) patients by inhibiting telomerase activity. Protein kinase, membrane associated tyrosine/threonine 1(Pkmyt1), a member of the WEE1 family, has been found to interact with MCRS1 by yeast two-hybrid assay; however, how these two proteins interact in GC is still unclear. Hence, this study aimed to investigate the effect of MCRS1 interaction with Pkmyt1 on GC cell proliferation, migration, and invasion. Initially, we observed increased expression of MCRS1 in GC SGC-7901 cells and decreased expression in GC BGC-823 cells. Hence, we down-regulated MCRS1 expression in SGC-7901 cells and up-regulated it in BGC-823 cells. Our results showed that overexpression of MCRS1 inhibits the growth, invasion and migration of GC cells, while downregulation of MCRS1 promotes the growth, invasion and migration of GC cells. When MK1775, an inhibitor of WEE1 kinase, was added after downregulation of MCRS1, phenotypic recovery effects were observed. Overexpression of MCRS1 also inhibited the expression of Pkmyt1 and vice versa. This indicated that there might be a possible interaction between MCRS1 and Pkmyt1. Furthermore, immunoprecipitation assay revealed the interaction between MCRS1 and Pkmyt1 in virto, and immunofluorescence experiments showed that the two proteins were co-localized in the cytoplasm. In conclusion, our study confirmed the specific tumor suppressive activity of MCRS1 in GC proliferation, invasion and migration and suggested that it might inhibit the progression of GC through its interaction with Pkmyt1.