Merck
  • A combined computational/experimental study on HSA binding of two water-soluble Schiff base ligands derived from pyridine derivative and ethylendiamine.
A combined computational/experimental study on HSA binding of two water-soluble Schiff base ligands derived from pyridine derivative and ethylendiamine.
Journal of biomolecular structure & dynamics (2018-02-03)
Hajar Molaee, Mehdi Sahihi, Majid Moghadam, Valiollah Mirkhani, Shahram Tangestaninejad, Iraj Mohammadpoor-Baltork, Hadi Amiri Rudbari