Merck
NanoSIMS imaging reveals unexpected heterogeneity in nutrient uptake by brown adipocytes.
Biochemical and biophysical research communications (2018-09-19)
Cuiwen He, Xuchen Hu, Thomas A Weston, Rachel S Jung, Patrick Heizer, Yiping Tu, Rochelle Ellison, Ken Matsumoto, Holger Gerhardt, Peter Tontonoz, Loren G Fong, Stephen G Young, Haibo Jiang
ZUSAMMENFASSUNG
Heterogeneity in the metabolic properties of adipocytes in white adipose tissue has been well documented. We sought to investigate metabolic heterogeneity in adipocytes of brown adipose tissue (BAT), focusing on heterogeneity in nutrient uptake. To explore the possibility of metabolic heterogeneity in brown adipocytes, we used nanoscale secondary ion mass spectrometry (NanoSIMS) to quantify uptake of lipids in adipocytes interscapular BAT and perivascular adipose tissue (PVAT) after an intravenous injection of triglyceride-rich lipoproteins (TRLs) containing [2H]triglycerides (2H-TRLs). The uptake of deuterated lipids into brown adipocytes was quantified by NanoSIMS. We also examined 13C enrichment in brown adipocytes after administering [13C]glucose or 13C-labeled mixed fatty acids by gastric gavage. The uptake of 2H-TRLs-derived lipids into brown adipocytes was heterogeneous, with 2H enrichment in adjacent adipocytes varying by more than fourfold. We also observed substantial heterogeneity in 13C enrichment in adjacent brown adipocytes after administering [13C]glucose or [13C]fatty acids by gastric gavage. The uptake of nutrients by adjacent brown adipocytes within a single depot is variable, suggesting that there is heterogeneity in the metabolic properties of brown adipocytes.