Merck
  • Integrin Mechano-chemical Signaling Generates Plasma Membrane Nanodomains that Promote Cell Spreading.
Integrin Mechano-chemical Signaling Generates Plasma Membrane Nanodomains that Promote Cell Spreading.
Cell (2019-05-21)
Joseph Mathew Kalappurakkal, Anupama Ambika Anilkumar, Chandrima Patra, Thomas S van Zanten, Michael P Sheetz, Satyajit Mayor
ZUSAMMENFASSUNG
Glycosylphosphatidylinositol-anchored proteins (GPI-APs) are a major class of lipid-anchored plasma membrane proteins. GPI-APs form nanoclusters generated by cortical acto-myosin activity. While our understanding of the physical principles governing this process is emerging, the molecular machinery and functional relevance of GPI-AP nanoclustering are unknown. Here, we first show that a membrane receptor signaling pathway directs nanocluster formation. Arg-Gly-Asp motif-containing ligands bound to the β1-integrin receptor activate src and focal adhesion kinases, resulting in RhoA signaling. This cascade triggers actin-nucleation via specific formins, which, along with myosin activity, drive the nanoclustering of membrane proteins with actin-binding domains. Concurrently, talin-mediated activation of the mechano-transducer vinculin is required for the coupling of the acto-myosin machinery to inner-leaflet lipids, thereby generating GPI-AP nanoclusters. Second, we show that these nanoclusters are functional; disruption of their formation either in GPI-anchor remodeling mutants or in vinculin mutants impairs cell spreading and migration, hallmarks of integrin function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Y-27632 -dihydrochlorid, ≥98% (HPLC)
Sigma-Aldrich
CK-666, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Aktin (20-33) in Kaninchen hergestellte Antikörper, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Integrin-β1-Antikörper, aktiviert, Klon HUTS-4, azidfrei, clone HUTS-4, Chemicon®, from mouse
Sigma-Aldrich
PP2, PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC50 = 4, 5, 5, &100 nM for p56lck, p59fynT, Hck, & Src, respectively).
Sigma-Aldrich
ML-7, powder
Sigma-Aldrich
SMIFH2, ≥98% (HPLC)
Sigma-Aldrich
PF-573228, ≥95% (HPLC)
Sigma-Aldrich
Anti-Talin antibody, Mouse monoclonal, clone 8D4, purified from hybridoma cell culture