Merck
Activated MLKL attenuates autophagy following its translocation to intracellular membranes.
Journal of cell science (2019-02-03)
Daniel Frank, David L Vaux, James M Murphy, James E Vince, Lisa M Lindqvist
ZUSAMMENFASSUNG
Necroptosis is an inflammatory form of programmed cell death mediated by the pseudokinase mixed-lineage kinase domain-like protein (MLKL). Upon phosphorylation by receptor-interacting protein kinase-3 (RIPK3), MLKL oligomerizes, and translocates to and disrupts the plasma membrane, thereby causing necroptotic cell lysis. Herein, we show that activation of necroptosis in mouse dermal fibroblasts (MDFs) and HT-29 human colorectal cancer cells results in accumulation of the autophagic marker, lipidated LC3B (also known as MAP1LC3B), in an MLKL-dependent manner. Unexpectedly, the necroptosis-induced increase in lipidated LC3B was due to inhibition of autophagic flux, not the activation of autophagy. Inhibition of autophagy by MLKL correlated with a decrease in autophagosome and/or autolysosome function, and required the association of activated MLKL with intracellular membranes. Collectively, our findings uncover an additional role for the MLKL pseudokinase, namely to inhibit autophagy during necroptosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-MLKL-Antikörper, Klon 3H1, clone 3H1, from rat