Merck
  • Polyphenolic profile and antioxidant activity of meristem and leaves from "chagual" (Puya chilensis Mol.), a salad from central Chile.
Polyphenolic profile and antioxidant activity of meristem and leaves from "chagual" (Puya chilensis Mol.), a salad from central Chile.
Food research international (Ottawa, Ont.) (2018-10-27)
Felipe Jiménez-Aspee, Cristina Theoduloz, Sergio Gómez-Alonso, Isidro Hermosín-Gutiérrez, Marisol Reyes, Guillermo Schmeda-Hirschmann
ZUSAMMENFASSUNG
The Bromeliaceae Puya chilensis Mol. is a native monocotyledonous food plant that can be found in central Chile. It is traditionally known as chagual. The tender basal part of the leaves, just starting from the meristem, are consumed as a salad. The aim of this work was to describe the phenolic content and composition of the meristem and leaves of chagual, as well as their antioxidant capacity and inhibitory activity against metabolic syndrome-associated enzymes. Samples of chagual, including two cultivated and three wild growing plants, were analyzed and compared for composition and bioactivity. From the phenolic enriched extract of the plant (PEE), 26 compounds were tentatively identified by HPLC-DAD-ESI-MSn, including 12 hydroxycinnamic acids and 14 flavonoids. The main compounds were identified as diferuloyl hexaric acid isomers and 5-p-Coumaroylquinic acid. The compounds were quantified in both meristem and leaves. The PEE content was up to ten times higher in the meristem than in the leaves, ranging from 0.18 to 124.08 mg/g PEE. The samples inhibited α-glucosidase, but did not show effect on α-amylase and pancreatic lipase. This is the first report on the polyphenol composition and bioactivity of the edible components of the chagual food plant.