Merck
  • Skeletal muscles of hibernating black bears show minimal atrophy and phenotype shifting despite prolonged physical inactivity and starvation.
Skeletal muscles of hibernating black bears show minimal atrophy and phenotype shifting despite prolonged physical inactivity and starvation.
PloS one (2019-04-19)
Mitsunori Miyazaki, Michito Shimozuru, Toshio Tsubota
ZUSAMMENFASSUNG
Hibernating mammals experience prolonged periods of torpor and starvation during winter for up to 5-7 months. Though physical inactivity and malnutrition generally lead to profound loss of muscle mass and metabolic dysfunction in humans, hibernating bears show limited muscle atrophy and can successfully maintain locomotive function. These physiological features in bears allow us to hypothesize that hibernating bears uniquely alter the regulation of protein and energy metabolism in skeletal muscle which then contributes to "muscle atrophy resistance" against continued physical inactivity. In this study, alteration of signaling pathways governing protein and energy metabolisms was examined in skeletal muscle of the Japanese black bear (Ursus thibetanus japonicus). Sartorius muscle samples were collected from bear legs during late November (pre-hibernation) and early April (post-hibernation). Protein degradation pathways, through a ubiquitin-proteasome system (as assessed by increased expression of murf1 mRNA) and an autophagy-dependent system (as assessed by increased expression of atg7, beclin1, and map1lc3 mRNAs), were significantly activated in skeletal muscle following hibernation. In contrast, as indicated by a significant increase in S6K1 phosphorylation, an activation state of mTOR (mammalian/mechanistic target of rapamycin), which functions as a central regulator of protein synthesis, increased in post-hibernation samples. Gene expression of myostatin, a negative regulator of skeletal muscle mass, was significantly decreased post-hibernation. We also confirmed that the phenotype shifted toward slow-oxidative muscle and mitochondrial biogenesis. These observations suggest that protein and energy metabolism may be altered in skeletal muscle of hibernating bears, which then may contribute to limited loss of muscle mass and efficient energy utilization.